missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, for HPLC, Stabilised with Amylene, Fisher Chemical™

CH2Cl2, CAS Number-75-09-2, 2.5L, 39 deg.C, Colorless, 84.93g/mol, -97 deg.C, 881, Amber glass bottle, Liquid, 2.93, 350mbar at 20 deg.C, 0.43 mPaS at 20 deg.C

Brand:  Fisher Chemical D/1856/PB17

49.60 EUR valid until 2021-02-28
Use promo code "14802" to get your promotional price.



Code : 25

Additional Details : CAS Number : 75-09-2 Weight : 3.31250kg Transport : UN number : 1593 Chem class : 6.1 Pack group : III


 View more versions of this product

Product Code. 10010120

  • 124.00 € / 2.50 L

Estimated Shipment
04-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CH2Cl2, CAS Number-75-09-2, 2.5L, 39 deg.C, Colorless, 84.93g/mol, -97 deg.C, 881, Amber glass bottle, Liquid, 2.93, 350mbar at 20 deg.C, 0.43 mPaS at 20 deg.C

Specifications

Specifications

75-09-2
881
Liquid
350mbar at 20°C
0.43 mPaS at 20°C
Colorless
2.5L
CH2Cl2
84.93g/mol
Plastic Coated Glass bottle
2.93
39°C
-97°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dichloromethane

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents