missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorvos 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Dichlorvos |
| CAS | 62-73-7 |
| Analyte or Component Names | Dichlorvos |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H7 Cl2 O4 P |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Phosphoric acid, 2,2-dichloroethenyl dimethyl ester, Phosphoric acid, 2,2-dichlorovinyl dimethyl ester (6CI,8CI), Ethenol, 2,2-dichloro-, dimethyl phosphate (8CI), 2,2-Dichloroethenyl dimethyl phosphate, 2,2-Dichlorovinyl dimethyl phosphate, Alatox, Atgard, Atgard V, BAY-b 4986, Bibesol, Brevinyl, Brevinyl E 50, Canogard, Chlorvinphos, DDVP, DDVP (insecticide), Dash, Dedevap, Des, Des (phosphate), Dichlofos, Dichlorman, Dichlorovinphos, Dichlorovos, Dichlorvos, Dimethyl 2,2-dichloroethenyl phosphate, Dimethyl 2,2-dichlorovinyl phosphate, Dimethyl dichlorovinyl phosphate, Divipan, ENT 20738, Equigard, Equigel, Estrosel, Estrosol, Estrosol (pesticide), Fecama, Fekama, Ferag ID TM, Insectigas D, Lava 100EC, Mopari, NSC 6738, Nefrafos, Nerkol, Nogos, Nogos 500, Nogos 50EC, Nogos G, Novotox, Nuvan, Nuvan 100EC, Nuvan 500 EC, Nuvan 7, Nuvan 76EC, Nuvos, O,O-Dimethyl 2,2-dichlorovinyl phosphate, OKO, OMS 14, Panaplate, Phosvit, Phosvit 75, Prima U, SD 1750, Szklarniak, TAP 9VP, Task, Task (insecticide), Unifos, Unifos (pesticide), Unifos 50EC, Unitox, Vapona, Vapona Insecticide, Vaponite, Vinylofos, Vinylophos, Winylophos |
| InChI Formula | InChI=1S/C4H7Cl2O4P/c1-8-11(7,9-2)10-3-4(5)6/h3H,1-2H3 |
| SMILES | COP(=O)(OC)OC=C(Cl)Cl |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction