Learn More
Diethoxymethane, 99%, pure, stabilized, ACROS Organics™
Brand: Acros Organics 296450050
Code : 29
Additional Details : CAS Number : 462-95-3 Weight : 0.05420kg Transport : UN number : 2373 Chem class : 3 Pack group : II
Packaging | Glass bottle |
---|---|
Quantity | 5mL |
Chemical Identifiers
462-95-3 | |
104.15 | |
KLKFAASOGCDTDT-UHFFFAOYSA-N | |
10024 | |
CCOCOCC |
C5H12O2 | |
MFCD00009246 | |
diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene | |
ethoxymethoxyethane |
Specifications
Diethoxymethane | |
−5°C | |
Glass bottle | |
0.831 | |
0.405 mPa.s (20°C) | |
87.5°C | |
-66.5°C | |
99% | |
98.5% min. (GC) | |
CH3CH2OCH2OCH2CH3 | |
diethoxymethane, ethylal, formaldehyde diethyl acetal, diethylformal, diethoxy methane, methane, diethoxy, ethoxymethoxy ethane, ethoxymethyl ethyl ether, 1,1-diethoxymethane, 3,5-dioxaheptene | |
KLKFAASOGCDTDT-UHFFFAOYSA-N | |
ethoxymethoxyethane | |
10024 | |
Liquid | |
Pure |
0.8310g/mL | |
Authentic | |
1.3720 to 1.3740 | |
100ppm BHT max. | |
0.02% max. (K.F.) | |
Colorless | |
5mL | |
462-95-3,128-37-0 | |
C5H12O2 | |
MFCD00009246 | |
Solubility in water: 4.2%. Other solubilities: miscible with ethanol, ethyl ether, very soluble in acetone, soluble in benzene, chloroform | |
CCOCOCC | |
104.15 | |
104.15 | |
99% |