missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ≥99%, for HPLC, stabilized with Ethanol, ACROS Organics™

Brand:  Acros Organics 410040025

Code : 29

Additional Details : CAS Number : 60-29-7 Weight : 3.28500kg Transport : UN number : 1155 Chem class : 3 Pack group : I

Packaging Glass bottle
Quantity 2.5L

 View more versions of this product

Product Code. 10021224

  • 120.88 € / 2.50 L

Estimated Shipment
01-07-2021

Please to purchase this item. Need a web account? Register with us today!


Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

For HPLC
Specifications

Chemical Identifiers

60-29-7
74.123
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC

Specifications

Diethyl ether
0.009 max. at 400nm, 0.022 max. at 280nm, 0.046 max. at 270nm, 0.097 max. at 250nm, 0.301 max. at 230nm, 0.523 max. at 210nm, 1 (1 cm cell vs water) at 208nm
0.0002 meq/g max.
C4H10O
MFCD00011646
11,3762
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
≥98%
0.7140g/mL
Glass bottle
1.3510 to 1.3530
0.714
0.05% max.
0.2448 mPa.s (20°C)
Undesignated
2.5L
Stabilized
60-29-7
0.0002 meq/g max.
(CH3CH2)2O
01, 314
15, 3861
Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
CCOCC
74.123
CHEBI:35702
Liquid
HPLC
0.0003% max.
1ppm max. (at lot release)
Solubility in water: 69g/L (20°C). Other solubilities: soluble in concentrated hydrochloric acid, miscible with lower aliphatic alcohols, benzene,, chloroform, petroleum ether and many oils
2% ethanol
−45°C
34.6°C
-116.0°C
Videos
SDS Documents