missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ethylphosphonate, 98%, ACROS Organics™

Brand:  Acros Organics 333690050

Code : 29

Additional Details : CAS Number : 78-38-6 Weight : 0.05500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10001781

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 01 885 5854  for assistance.


Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

Diethyl ethylphosphonate
≥97.5%
C6H15O3P
78-38-6
98%
MFCD00039888

Specifications

Diethyl ethylphosphonate
Clear colorless
78-38-6
Glass bottle
Liquid
98%
MFCD00039888
CCOP(=O)(CC)OCC
166.157
166.16
5g
Authentic
1.4155 to 1.4175 (20°C, 589nm)
≥97.5%
C6H15O3P
AATNZNJRDOVKDD-UHFFFAOYSA-N
1-[ethoxy(ethyl)phosphoryl]oxyethane
6534
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diethyl ethylphosphonate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS