Learn More
Diethyl methylphosphonate, 96%, ACROS Organics™
Brand: Acros Organics 333680250
Code : 29
Additional Details : CAS Number : 683-08-9 Weight : 0.07500kg
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Description
For C-C Bond Formation (Olefination)Chemical Identifiers
683-08-9 | |
152.13 | |
NYYLZXREFNYPKB-UHFFFAOYSA-N | |
12685 | |
CCOP(=O)(C)OCC |
C5H13O3P | |
MFCD00009813 | |
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h | |
1-[ethoxy(methyl)phosphoryl]oxyethane |
Specifications
Diethyl methylphosphonate | |
Authentic | |
1.4130 to 1.4150 | |
Colorless to Yellow | |
96% | |
95% min. (GC) | |
MFCD00009813 | |
NYYLZXREFNYPKB-UHFFFAOYSA-N | |
1-[ethoxy(methyl)phosphoryl]oxyethane | |
12685 | |
Liquid |
75°C | |
Glass bottle | |
78.0°C to 81.0°C (15.0mmHg) | |
25g | |
683-08-9 | |
C5H13O3P | |
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h | |
CCOP(=O)(C)OCC | |
152.13 | |
152.13 | |
96% |