missing translation for 'onlineSavingsMsg'
Learn More

Diethyl methylphosphonate, 96%, Thermo Scientific Chemicals

Catalog Number 10612992
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
683-08-9
C5H13O3P
152.13
MFCD00009813
NYYLZXREFNYPKB-UHFFFAOYSA-N
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h
12685
1-[ethoxy(methyl)phosphoryl]oxyethane
CCOP(C)(=O)OCC
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
683-08-9
C5H13O3P
152.13
MFCD00009813
NYYLZXREFNYPKB-UHFFFAOYSA-N
diethyl methylphosphonate, diethyl methanephosphonate, phosphonic acid, methyl-, diethyl ester, demp, 1-ethoxy methyl phosphoryl oxyethane, methyl-phosphonicacidiethylester, diethylmethylphosphonate, diethyl methyphosphonate, diethoxymethylphosphine oxide, acmc-1b13h
12685
1-[ethoxy(methyl)phosphoryl]oxyethane
CCOP(C)(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 78.0°C to 81.0°C (15.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4130 to 1.4150
Quantity 5 g
Formula Weight 152.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Diethyl methylphosphonate, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'