missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (hydroxymethyl)phosphonate, 97%, Thermo Scientific™

Brand:  Thermo Scientific Acros 320950050

Code : 29

Additional Details : CAS Number : 3084-40-0 Weight : 0.02444kg

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10003890

Quantity Price
1 51.80 € / 5g
10 47.94 € / 5g
Estimated Shipment: 02-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

3084-40-0
168.13
RWIGWWBLTJLKMK-UHFFFAOYSA-N
76513
CCOP(=O)(CO)OCC
C5H13O4P
MFCD00014418
diethyl hydroxymethyl phosphonate, diethyl hydroxymethylphosphonate, phosphonic acid, hydroxymethyl-, diethyl ester, hydroxymethyl phosphonic acid, diethyl ester, hydroxymethyl phosphonic acid diethyl ester, phosphonic acid, p-hydroxymethyl-, diethyl ester, acmc-1corl, diethoxyphosphoryl-methanol
diethoxyphosphorylmethanol

Specifications

Diethyl (hydroxymethyl)phosphonate
>110°C
Glass bottle
1.14
Colorless to Yellow
3084-40-0
C5H13O4P
MFCD00014418
RWIGWWBLTJLKMK-UHFFFAOYSA-N
diethoxyphosphorylmethanol
76513
Viscous Liquid
1.1400g/mL
Authentic
1.4370 to 1.4410
124.0°C to 126.0°C (3.0 mmHg)
5g
96% min. (GC)
HOCH2P(O)(OC2H5)2
diethyl hydroxymethyl phosphonate, diethyl hydroxymethylphosphonate, phosphonic acid, hydroxymethyl-, diethyl ester, hydroxymethyl phosphonic acid, diethyl ester, hydroxymethyl phosphonic acid diethyl ester, phosphonic acid, p-hydroxymethyl-, diethyl ester, acmc-1corl, diethoxyphosphoryl-methanol
CCOP(=O)(CO)OCC
168.13
168.13
97%
Videos
SDS Documents
Certificates

RUO – Research Use Only