missing translation for 'onlineSavingsMsg'
Learn More

Diethylzinc, 1.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 377311000

Code : 29

Additional Details : CAS Number : 557-20-0 Weight : 0.09130kg Transport : UN number : 3399 Chem class : 4.3 Pack group : I


 View more versions of this product

Product Code. 10464542

  • 146.00 € / 100mL
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethylzinc
1.5M Solution in Toluene
C4H10Zn
MFCD00009021
0.9130g/mL
diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn
HQWPLXHWEZZGKY-UHFFFAOYSA-N
diethylzinc
11185
AcroSeal™ Glass bottle
100 mL
Colorless
557-20-0,108-88-3
(C2H5)2Zn
1.4 to 1.6M
15, 3151
Solubility in water: reacts violently. Other solubilities: miscible with most hydrocarbons
CC[Zn]CC
123.50
123.5
0.913
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Diethylzinc, solution in toluene

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

Supplemental information
  • MIXTURE LIST-Contains : Toluene, Diethylzinc

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only