Learn More
BD DIFF-SAFE™ Blood Dispenser
Used to prepare blood smear slides directly from a Vacutainer™ blood collection tube without taking off the cap. BD DIFF-SAFE™ Blood Dispensers are inserted into a Hemogard™ closure and pressed against a microsoft slide to deliver a blood drop without the risk of spillage.
Brand: BD 366005
Code : 02
Additional Details : Weight : 5.10000kg
Description
The BD Vacutainer™ system is a closed system optimally designed for controlling pre-analytical variability throughout the specimen collection and handling process. The DIFF-SAFE™ blood dispenser prepares blood slides from a blood collection tube without removing the stopper from the tube.- Features a non-sterile cannula that inserts easily through the rubber stopper of a blood collection tube
- Invert the tube and press the dispenser against a slide to dispense a drop to prepare a blood smear
- Compatible with BD Vacutainer™ tubes with Hemogard™ closures
- Limits exposure to blood samples, which may contain bloodborn pathogens
- Keeps closure in place to prevent spills, leaks and contamination of samples
Specifications
White | |
Yes | |
Cannula |
Preparing blood smear slides from a blood collection tube. | |
Vacutainer Blood Collection Tubes with Hemogard Closures | |
Non-sterile |