missing translation for 'onlineSavingsMsg'
Learn More

Diiodomethane, 99+%, stabilized with silver wire, Thermo Scientific Chemicals

Catalog Number p-4510514
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
75-11-6, 7440-22-4
CH2I2
267.84
MFCD00001079
NZZFYRREKKOMAT-UHFFFAOYSA-N
methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
6346
diiodomethane
ICI
75-11-6, 7440-22-4
CH2I2
267.84
MFCD00001079
NZZFYRREKKOMAT-UHFFFAOYSA-N
methylene iodide, methane, diiodo, methylene diiodide, mi-gee, dijodmethan, methylenjodid, dijodmethan czech, methylenjodid czech, di-iodomethane, diiodo-methane
6346
diiodomethane
ICI

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 6.0°C
Color Red to Yellow
Density 3.3200g/mL
Boiling Point 181.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH2I2
Refractive Index 1.737
Quantity 25 g
Beilstein 01,71
Fieser 09,170; 11,182; 17,116
Merck Index 15,6137
Specific Gravity 3.32
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,propanol,isopropanol,,hexane,cyclohexane,ether,chloroform and benzene
Stabilizer Silver wire
Viscosity 2.80 mPa.s (20°C)
Formula Weight 267.83
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diiodomethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diiodomethane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dijodmethan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'