Learn More
Diisobutylaluminium hydride, 1M solution in hexane, AcroSeal™, ACROS Organics™
Brand: Acros Organics 183794000
Code : 29
Additional Details : CAS Number : 1191-15-7 Weight : 0.33040kg Transport : UN number : 3394 Chem class : 4.2 Pack group : I
Packaging | AcroSeal™ Glass Bottle |
---|---|
Quantity | 400mL |
Description
Lewis acid, Reduction reagentChemical Identifiers
1191-15-7 | |
142.22 | |
DIBAL-H |
C8H19Al | |
MFCD00008928 |
Specifications
Diisobutylaluminium hydride | |
−23°C | |
0.701 | |
-70.0°C | |
1M Solution in Hexane | |
C8H19Al | |
MFCD00008928 | |
01,260; 02,140; 03,101; 04,158; 05,224; 06,198; 07,111; 08,173; 09,171; 10,149; 11,185; 12,191; 13,115; 14,138; 15,137; 16,27; 17,123 | |
DIBAL-H | |
142.22 | |
Solution |
0.7010g/mL | |
AcroSeal™ Glass Bottle | |
Undesignated | |
400mL | |
1191-15-7,110-54-3 | |
[(CH3)2CHCH2]2AlH | |
04, IV, 4400 | |
15, 3212 | |
Solubility in water: reacts | |
142.22 |
Safety and Handling
- Diisobutylaluminium hydride, 1M solution in hexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
- Pyrophoric liquid Category 1
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 1 A
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H361f-Suspected of damaging fertility.
- EUH014-Reacts violently with water.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P231+P232-Handle under inert gas. Protect from moisture.
- P302+P334-IF ON SKIN: Immerse in cool water/wrap in wet bandages.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P273-Avoid release to the environment.
- P402+P404-Store in a dry place. Store in a closed container.
- P222-Do not allow contact with air.
Supplemental information
- MIXTURE LIST-Contains : Hexane, Diisobutylaluminum hydride