missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dilithium tetrabromonickelate(II), 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
170.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Blue-Green to Brown |
Physical Form | Solution |
Quantity | 800 mL |
Linear Formula | Li2NiBr4 |
Flash Point | −18°C |
Formula Weight | 392.19 |
Packaging | AcroSeal™ Glass Bottle |
Specific Gravity | 1.065 |
Density | 1.0650g/mL |
Chemical Name or Material | Dilithium tetrabromonickelate(II) |
Safety and Handling
Product Identifier
- Dilithium tetrabromonickelate(II), 0.5M solution in THF
- Danger
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H350-May cause cancer.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'