Learn More
Dimethylamine, 2M solution in methyl alcohol, AcroSeal™, ACROS Organics™
Brand: Acros Organics 370261000
Code : 29
Additional Details : CAS Number : 124-40-3 Weight : 0.12750kg Transport : UN number : 3286 Chem class : 3 Pack group : II
Packaging | AcroSeal™ Glass bottle |
---|---|
Quantity | 100mL |
Chemical Identifiers
124-40-3 | |
45.08 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
CHEBI:17170 | |
CNC |
C2H7N | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
674 | |
N-methylmethanamine |
Specifications
Dimethylamine | |
5°C | |
0.775 | |
124-40-3,67-56-1 | |
NH(CH3)2 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
674 | |
45.08 |
0.7750g/mL | |
AcroSeal™ Glass bottle | |
100mL | |
C2H7N | |
15,325 | |
Solubility in water: soluble. Other solubilities: soluble in most organic solvents | |
CNC | |
45.08 | |
CHEBI:17170 |
Safety and Handling
- Dimethylamine, 2M solution in methyl alcohol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.