missing translation for 'onlineSavingsMsg'
Learn More

Dinitrodurene, 97%, Thermo Scientific™

Catalog Number 15269616
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
5465-13-4
AEPQXGFMAZTUEA-UHFFFAOYSA-N
1,2,4,5-tetramethyl-3,6-dinitro-benzene, dinitrotrodurene, 3,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethyl-p-dinitrobenzene, 1,4,5-tetramethyl-3,6-dinitrobenzene, benzene,2,4,5-tetramethyl-3,6-dinitro, 1,4-dinitro-2,3,5,6-tetramethylbenzene 97+%
79593
1,2,4,5-tetramethyl-3,6-dinitrobenzene
CC1=C(C(=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-])C
This item is not returnable. View return policy
5465-13-4
AEPQXGFMAZTUEA-UHFFFAOYSA-N
1,2,4,5-tetramethyl-3,6-dinitro-benzene, dinitrotrodurene, 3,2,4,5-tetramethylbenzene, 2,3,5,6-tetramethyl-p-dinitrobenzene, 1,4,5-tetramethyl-3,6-dinitrobenzene, benzene,2,4,5-tetramethyl-3,6-dinitro, 1,4-dinitro-2,3,5,6-tetramethylbenzene 97+%
79593
1,2,4,5-tetramethyl-3,6-dinitrobenzene
CC1=C(C(=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-])C
Melting Point 208.0°C to 213.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C10H12N2O4
Linear Formula (CH3)4C6(NO2)2
Quantity 25 g
Molecular Weight (g/mol) 224.216
Formula Weight 224.22
Percent Purity 97%
Chemical Name or Material Dinitrodurene
compliance-icons compliance-icons
Product Identifier
  • Dinitrodurene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'