missing translation for 'onlineSavingsMsg'
Learn More

Dinonyl phthalate, 96%, mixture of isomers, ACROS Organics™

Brand:  Acros Organics 408771000

Code : 29

Additional Details : CAS Number : 84-76-4 Weight : 0.15000kg

Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 11945041

  • 280.00 € / 100g

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

84-76-4
418.62
DROMNWUQASBTFM-UHFFFAOYSA-N
6787
CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC
C26H42O4
MFCD00036237
dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
dinonyl benzene-1,2-dicarboxylate

Specifications

Dinonyl phthalate
216°C
Glass bottle
0.97
279.0°C to 287.0°C
100g
95% min. (sum of isomers) (GC)
MFCD00036237
Solubility in water: <1g/L (20°)
CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC
418.62
418.61
96%
0.9700g/mL
Authentic
1.4825 to 1.4845
166 mPa.s (20°C)
Yellow to Brown
84-76-4
C26H42O4
dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
DROMNWUQASBTFM-UHFFFAOYSA-N
dinonyl benzene-1,2-dicarboxylate
6787
Oily Liquid
Videos
SDS Documents
Certificates