missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipentyl phthalate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -55.0°C |
Color | Colorless to Yellow |
Density | 1.0230g/mL |
Boiling Point | 342.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (C(O)OC5H11)2C6H4 |
Refractive Index | 1.4885 to 1.4905 |
Quantity | 500 g |
Show More |
Safety and Handling
Product Identifier
- Dipentyl phthalate
- Danger
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1B
- H400-Very toxic to aquatic life.
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'