missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Discharge Tubes for Dispensette™ S Bottletop Dispensers

For use with Dispensette S Bottletop Dispensers.

Brand:  BRAND™ 708112

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 15549724

  • 103.00 € / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket

Description

Description

  • Fine tip
  • Volume: 5 and 10mL
  • Available with or without recirculation valve
Specifications

Specifications

Discharge Tube
Dispensette S Bottletop Dispensers
Fine tip
5, 10 mL
Recirculation valve
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers