Learn More
Kimberly-Clark Professional™ Facial Tissue Dispenser
Promotes hygiene and helps control costs. Kimberly-Clark Professional™ Facial Tissue Dispenser improves staff and customer comfort and care.
Brand: Kimberly-Clark Professional™ 7820
33.61 EUR valid until 2024-12-31
Use promo code "22916" to get your promotional price.
Code : 02
Additional Details : Weight : 0.39600kg
Description
- Quick-fill, hassle-free maintenance
- Wall-mounted
- Chromed metallic silver finish
- Ideal for dispensing facial tissues in office areas and hotel washrooms that maintain a luxury image
Specifications
Facial Tissue Dispenser | |
282 x 153 x 84 mm | |
28.2 cm | |
1 Ea. |
Silver | |
11 in. | |
Plastic |