missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Dispensette™ S Fixed-volume Bottletop Dispensers
The Dispensette™ S Fixed Volume Bottletop Dispensers feature a “floating piston” design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.
400.00 € - 504.00 €
Specifications
Product Type | Bottletop Dispenser |
---|---|
Autoclavable | Autoclavable at 121°C |
Certifications/Compliance | ISO 9001 |
Color | Red |
For Use With (Application) | Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents |
Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual
Product Code | Brand | Volume (Metric) | Accuracy | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Accuracy | Description | Price | Quantity & Availability | |||
15458053
|
BRAND™
4600211 |
1 mL | 0.6% (6μL) | With recirculation valve |
504.00 €
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||
15478053
|
BRAND™
4600221 |
2 mL | 0.5% (10μL) | With recirculation valve |
489.00 €
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||
15498053
|
BRAND™
4600231 |
5 mL | 0.5% (25μL) | With recirculation valve |
489.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
15438063
|
BRAND™
4600241 |
10 mL | 0.5% (50μL) | With recirculation valve |
504.00 €
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||
15448053
|
BRAND™
4600210 |
1 mL | 0.6% (6μL) | Without recirculation valve |
425.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
15488053
|
BRAND™
4600230 |
5 mL | 0.5% (25μL) | Without recirculation valve |
425.00 €
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||
15468053
|
BRAND™
4600220 |
2 mL | 0.5% (10μL) | Without recirculation valve |
425.00 €
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||
15408063
|
BRAND™
4600240 |
10 mL | 0.5% (50μL) | Without recirculation valve |
400.00 €
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||
Description
- New discharge tube, with or without recirculation valve
- New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
- Simple priming by optimized flow channels
- Less dispensing force - especially with large volume instruments
- Safe volume setting with internal scalloped track (analog-adjustable instruments)
- Pivoted screw cap - does not disturb dispensing
- Easy to calibrate and adjust in order to comply with GLP guidelines
- Easy to dismantle for cleaning
- Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
- 360° rotatable valve block allows bottle label to face user
- Telescoping filling tube adjusts easily to different size bottles
- Digital and analog-adjustable models
- DE-M marking
Warranty
One year
Specifications
Bottletop Dispenser | |
ISO 9001 | |
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents |
Autoclavable at 121°C | |
Red |