Learn More
Sartorius™ Disposable Chute Bags
Disposable waste removal bags for safe disposal of toxic and high potency samples handled in SWC Safety Weighing Cabinets. (Pack of 100)
Brand:  Sartorius™ YWCG02
Code : NEW
Additional Details : Weight : 0.99000kg
Product Code. 10549618
  • 108.00 € / Pack of 50
Estimated Shipment: 30-04-2024
to see stock.
Description
Description
Safety weighing cabinets ensure a safe working environment for laboratory staff without sacrificing ergonomics or precision in weighing of toxic and high-potency samples.
  • Designed for use with SWC Safety Weighing Cabinets
  • Disposal chute attaches securely to the side of the cabinet
  • Disposable chute bags enable safe disposal of substances without exposure to the lab
  • Contaminated materials can be disposed of safely in a double-bag system
  • Packaged: 100/Pack
Specifications
Specifications
YWCG02 Disposal Chute for Waste, SWC Safety Weighing Cabinet
Waste disposal
SWC
Transparent
100/Pk.