Learn More
DWK Life Sciences Disposable Culture Tube, from soda-lime glass, straight rim
Culture tubes are used for growing cultures from liquid media.
13.65 € - 79.45 €
Specifications
Material | Soda Lime Glass |
---|---|
For Use With | Growth of microorganisms; For microbiological work |
Autoclavable | Not Autoclavable |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15601604
|
DWK Life Sciences
231720862 |
6 mL | 11.75 mm |
14.35 €
Pack of 250 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
15213035
|
DWK Life Sciences
231721283 |
18 mL | 12.25 mm |
35.60 €
Pack of 250 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
15611604
|
DWK Life Sciences
231721883 |
8 mL | 10 mm |
30.10 €
Pack of 250 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
10595272
|
DWK Life Sciences
231720784 |
8 mL | 11.75 mm |
79.45 €
Pack of 550 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
16272261
|
DWK Life Sciences
231721986 |
19 mL | 15.5 mm |
30.90 €
Pack of 250 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
15299574
|
DWK Life Sciences
231721489 |
14 mL | 15.5 mm |
28.00 €
Pack of 250 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
15267556
|
DWK Life Sciences
231720365 |
4 mL | 10 mm |
13.65 €
Pack of 250 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
10598411
|
DWK Life Sciences
231720965 |
7 mL | 12.25 mm |
29.00 €
Pack of 500 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||||
10716961
|
DWK Life Sciences
231720998 |
6 mL | 12.25 mm |
40.15 €
Pack of 500 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
10766771
|
DWK Life Sciences
231721197 |
9 mL | 12.25 mm |
44.65 €
Pack of 500 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
15473520
|
DWK Life Sciences
231722193 |
22 mL | 15.5 mm |
39.05 €
Pack of 250 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
10060371
|
DWK Life Sciences
231721094 |
15 mL | 15.75 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Descripción
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today''s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. In addition to culture tubes made of DURAN™, our product range also includes culture tubes made of soda-lime glass. This is a glass of the third water resistance Class And belongs to the category of soda-lime glasses with a high proportion of alkali and alkaline earth oxides.
- Culture tubes with straight edges can be closed with metal caps or cotton plugs to allow air exchange
- The straight rim permits the use of Kapsenberg and metal caps
Especificaciones
Soda Lime Glass | |
Growth of microorganisms; For microbiological work | |
Not Autoclavable |