Learn More
Brand™ Blaubrand™ IntraMARK™ Disposable Micropipets
Constructed of disposable glass and conforms to DIN ISO 7550 standards. BRAND™ Blaubrand™ IntraMARK™ Disposable Micropipets include ring mark, and are CE-marked according to IVD Directive 98/79 CE. All surfaces are fire polished for smoothness and to minimize pipet adapter wear. Color-coded to ISO for clear identification.
Brand:  Brand™ 708722
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15212599
  • 131.00 € / Pack of 1000
In Stock
to see stock.
Add to basket
Description
Description
  • With ring mark
  • Conform to DIN ISO 7550
  • Calibrated to contain (TC, In.)
  • CE-marked according to IVD Directive 98/79 CE.
  • Fire polished surfaces inside and out are particulary smooth
  • The aspiration end is also fire polished to minimize pipet adapter wear.
  • Color code to ISO for clear identification.
Specifications
Specifications
≤±0.25%
DIN ISO 7550, IVD-Directive 98/79 CE-marked
Glass
250/Box
125 mm
To Contain
Marks at 25μL
Ring mark
White
Single-use