missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable PES Bottle Top Filters

Product Code. p-7113062 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
150 mL
500 mL
1000 mL
Pore Size:
0.20 μm
0.45 μm
Diameter (Metric) Membrane:
50 mm
75 mm
91 mm
Unit Size:
Pack of 12
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Pore Size Diameter (Metric) Membrane unitSize
15953307 150 mL 0.20 μm 50 mm Pack of 12
15963307 150 mL 0.45 μm 50 mm Pack of 12
15973307 500 mL 0.20 μm 75 mm Pack of 12
15983307 500 mL 0.45 μm 75 mm Pack of 12
15993307 1000 mL 0.20 μm 91 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Product Code. 15953307
Only null left
Add to basket
Edge
Add to basket
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.

Increase sample protection with Fisherbrand Sterile Disposable Bottle Top Filters with PES Membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.

  • PES membrane is low protein binding to maintain protein balance
  • PES membrane has low extractables to maintain media purity
  • Marked with catalog number, membrane type, pore size, lot number and expiration date for easy traceability
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
  • Bottom screw threads for attachment to bottles with 45 mm neck sizes
  • 0.2μm units for sterile filtration
  • 0.45μm units for fluid clarification and particle removal

Certifications

Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Non-pyrogenic per USP Standard Lot-to-Lot
Disposable Single-use
Material (Membrane) PES
Pore Size 0.20 μm
Sterility Sterile
Type Bottle Top Unit
Volume (Metric) 150 mL
Diameter (Metric) Membrane 50 mm

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.