Learn More
Description
Includes
Individually packaged sterile screw top style bottle cap
Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
- PES membrane is low protein binding to maintain protein balance
- PES membrane has low extractables to maintain media purity
- Upper chamber of filter units is clearly marked with pore size, membrane type, catalog number, lot number and expiration date
- Non-pyrogenic and non-cytotoxic
- Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
- 0.2μm units for sterile filtration
- 0.45μm units for fluid clarification and particle removal
Certifications
Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility
Specifications
Specifications
| Binding Capacity | Low binding |
| Certifications/Compliance | Non-pyrogenic per USP Standard Lot-to-Lot |
| Disposable | Single-use |
| Material | PES |
| Material (Membrane) | PES |
| Pore Size | 0.20 μm |
| Sterility | Sterile |
| For Use With (Application) | Sterile Filtration of media and serum |
| Type | Filter Unit |
| Volume (Metric) | 150 mL |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
