Learn More
Palintest™ Photometer Tube Test
Predispensed reagent tests require minimal handling and speed up processing. Palintest™; Photometer Tube Test reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.
Brand:  Palintest™ PL 503
Code : 04
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11988987
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • For ultimate convenience, the Palintest™ range of Tubetest reagents offer pre-dispensed reagents for minimal handling with barcoded reagent labelling for ease of use in wastewater applications.
  • For a complete wastewater analysis system, use Palintest™ tubetests with any Palintest™ multiparameter photometer and heater block.
  • Tubetest reagents are supplied in 16mm tubes for improved resolution, with greater sensitivity in the lower part of the measurement range.
Specifications
Specifications
Dissolved Oxygen
30
0 to 2.0mg/L
Photometer, heater block
1 Each
Photometer tube test reagent