missing translation for 'onlineSavingsMsg'
Learn More

Dithizone, ACS, 85+%, Thermo Scientific Chemicals

Catalog Number p-7050012
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
60-10-6
C13H12N4S
256.33
MFCD00003025
UOFGSWVZMUXXIY-UHFFFAOYSA-N
dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone
657262
1-anilino-3-phenyliminothiourea
S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
This item is not returnable. View return policy
60-10-6
C13H12N4S
256.33
MFCD00003025
UOFGSWVZMUXXIY-UHFFFAOYSA-N
dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone
657262
1-anilino-3-phenyliminothiourea
S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1

Dithizone is used as an indicator for minute amounts of lead. It acts as a chelating agent for heavy metal poisoning. As a ligand, it forms complexes with metals like zinc, lead and mercury. Further, it is used to assess the purity of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes. In addition to this, it is used as a titrant in the quantitative determination of heavy metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 168°C (decomposition)
Odor Odorless
Quantity 2 g
Beilstein 748838
Merck Index 14,3377
Solubility Information Soluble in ethanol,chloroform,dimethyl sulfoxide and pyridine. Insoluble in water.
Formula Weight 256.33
Percent Purity ≥85%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Dithizone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'