missing translation for 'onlineSavingsMsg'
Learn More

DL-1-Amino-2-propanol, 94%, contains approx. 5% 2-Amino-1-propanol, Thermo Scientific Chemicals

Catalog Number 10021500
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
78-96-6
C3H9NO
75.11
MFCD00008139
HXKKHQJGJAFBHI-UHFFFAOYNA-N
1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine
4
CHEBI:19030
1-aminopropan-2-ol
CC(O)CN
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
78-96-6
C3H9NO
75.11
MFCD00008139
HXKKHQJGJAFBHI-UHFFFAOYNA-N
1-amino-2-propanol, isopropanolamine, monoisopropanolamine, threamine, 2-propanol, 1-amino, 2-hydroxypropylamine, amino-2-propanol, mipa, 1-methyl-2-aminoethanol, 2-hydroxypropanamine
4
CHEBI:19030
1-aminopropan-2-ol
CC(O)CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -2°C
Density 0.9600g/mL
Boiling Point 161°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 93% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2NH2
Refractive Index 1.4470 to 1.449
Quantity 2.5 kg
Beilstein 04, 289
Specific Gravity 0.96
Solubility Information Solubility in water: freely soluble
Viscosity 31 mPa.s (20°C)
Formula Weight 75.11
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material DL-1-Amino-2-propanol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • DL-1-Amino-2-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'