missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Aminoadipic acid, 99%, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4258742
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
542-32-5
C6H11NO4
161.16
MFCD00063119,MFCD00063118
OYIFNHCXNCRBQI-UHFFFAOYNA-N
dl-2-aminoadipic acid, aminoadipic acid, alpha-aminoadipic acid, 2-aminoadipic acid, dl-alpha-aminoadipic acid, 2-aminoadipate, alpha-aminoadipate, hexanedioic acid, 2-amino, 2-amino-hexanedioic acid, dl-2-aminohexanedioic acid
469
CHEBI:37024
2-aminohexanedioic acid
NC(CCCC(O)=O)C(O)=O
This item is not returnable. View return policy
542-32-5
C6H11NO4
161.16
MFCD00063119,MFCD00063118
OYIFNHCXNCRBQI-UHFFFAOYNA-N
dl-2-aminoadipic acid, aminoadipic acid, alpha-aminoadipic acid, 2-aminoadipic acid, dl-alpha-aminoadipic acid, 2-aminoadipate, alpha-aminoadipate, hexanedioic acid, 2-amino, 2-amino-hexanedioic acid, dl-2-aminohexanedioic acid
469
CHEBI:37024
2-aminohexanedioic acid
NC(CCCC(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 176.0°C to 177.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,41
Solubility Information Solubility in water: .. Other solubilities: sparingly soluble in alcohol and ether
Formula Weight 161.16
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material DL-2-Aminoadipic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'