missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DNA Methyltransferase Inhibitor
Click to view available options
Quantity:
10 mg
Description
A cell-permeable, specific DNA methyltransferases inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 10 mg |
Description | Yellow |
For Use With (Application) | A cell-permeable, specific DNA methyltransferases inhibitor |
CAS | 48208-26-0 |
Molecular Formula | C19H14N2O4, C19H14N2O4 |
Formula Weight | 334.32g/mol |
Synonym | RG108; N-Phthalyl-L-tryptophan |
Sensitivity | Light Sensitive |
Product Type | Signal Transduction Reagent |
Form | Solid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'