missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3A / DNA Methyltransferase 3 Alpha Mouse, Clone: PCRP-DNMT3A-1E2, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1788-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
DNMT3A / DNA Methyltransferase 3 Alpha | |
Nuclear | |
PCRP-DNMT3A-1E2 | |
Unconjugated | |
Mouse | |
102kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
1788 | |
IgG1 |
Immunoprecipitation, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human DNMT3A protein | |
HeLa cells orPlacenta | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |