Learn More
Dodecanal, 95%, stab.
Description
Dodecyl aldehyde is used as a fragrance agent. It is used as a common ingredient in perfumery. It's end applications include soap, detergent, beauty care product, household product.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dodecyl aldehyde is used as a fragrance agent. It is used as a common ingredient in perfumery. It′s end applications include soap, detergent, beauty care product, household product.
Solubility
Soluble in chloroform, methanol, alcohol, and propylene glycol. Insoluble in glycerol and water.
Notes
Air sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container.
Specifications
Specifications
| Melting Point | 12°C |
| Density | 0.835 |
| Boiling Point | 184°C to 186°C (100 mmHg) |
| Flash Point | 101°C (214°F) |
| Assay Percent Range | 95% |
| Refractive Index | 1.435 |
| Quantity | 500 g |
| Beilstein | 1703917 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in chloroform,methanol,alcohol,and propylene glycol. Insoluble in glycerol and water. |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.