missing translation for 'onlineSavingsMsg'
Learn More

Manutan Dolly, steel, 300kg capacity

Brand:  Manutan A047446

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12877828

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Steel dolly with all round PVC buffer strip to protect walls and doorways. Non-slip PVC deck surface.
  • Dimensions (H x W x L): 135mm x 510mm x 870mm
  • Fitted with two fixed and two swivel castors
  • Load capacity: 300kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers