missing translation for 'onlineSavingsMsg'
Learn More

KARTELL DOSIPET S

Dosipet S is a series of adjustable volume repetitive dosing devices. They are made of high quality materials ensuring considerable mechanical and chemical resistance.

Brand:  KARTELL 8307000

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10020263

  • 497.00 € / Each
Estimated Shipment: 11-10-2024
to see stock.
Add to basket

Description

Description

  • Useful for dosing concentrated acids, polar and no-polar solvents.
  • High quality components and assembly make this series the ideal choice for measuring out organic polar and non- polar solvents, including halogenated solvents, and concentrated mineral acids, including particularly volatile acids such as nitric and trifluoroacetic acids.
  • In addition, they can be used with peroxides, hydrochloric acid and sulphuric acid.
  • It comes with a borosilicate glass piston.
  • The reagents come into contact only with parts made of PTFE
  • Precise and quick volume adjustment using the analogue slider
  • Precise graduated scale to ensure reproducibility of the dispensed volumes
  • No loss of reagent when eliminating air or filling
  • Dispensing without bubbles forming
  • The piston completely transfers the liquid with every movement
  • Total recovery of the liquid in the discharge tube by turning the delivery tip 180°
  • Autoclavable at +121°C for 20 minutes
Specifications

Specifications

± 0.06 mL
Autoclavable
White
Borosilicate Glass, PTFE
Analog
0.34 oz.
Dosipet S
None
1
10 mL
0.20 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers