Learn More
Dowex Marathon C, H+ form, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar B25721.0E
Code : NEW
Additional Details : Weight : 2.50000kg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is designed for demineralization using HCl or H2 SO4 as regenerate in co-current or counter-current operation.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Spezifikation
Dowex Marathon C | |
150604-77-6 | |
1.2 | |
Insoluble in water. |
H+ form | |
MFCD00212649 | |
2500 g |