Learn More
Doxepin hydrochloride, Thermo Scientific™
Potent H1-histamine receptor antagonist and a 5-HT receptor antagonist
46.50 € - 559.00 €
Chemical Identifiers
| CAS | 1229-29-4 |
|---|---|
| Molecular Formula | C19H22ClNO |
| Molecular Weight (g/mol) | 315.841 |
| MDL Number | MFCD00079135 |
| InChI Key | MHNSPTUQQIYJOT-SJDTYFKWSA-N |
| Synonym | doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride |
| PubChem CID | 6419921 |
| IUPAC Name | (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride |
| SMILES | CN(C)CCC=C1C2=CC=CC=C2COC3=CC=CC=C31.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15494919
|
Thermo Scientific Alfa Aesar
J62579.03 |
1g |
46.50 €
1g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15404929
|
Thermo Scientific Alfa Aesar
J62579.06 |
5g |
144.00 €
5g |
Estimated Shipment: 30-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15414929
|
Thermo Scientific Alfa Aesar
J62579.14 |
25g |
559.00 €
25g |
Estimated Shipment: 23-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water (100 mM), chloroform (1:2), alcohol (1:1), methanol, and DMSO.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| C19H22ClNO | |
| MFCD00079135 | |
| doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride | |
| (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride |
Specifications
| 1229-29-4 | |
| MFCD00079135 | |
| 14,3435 | |
| Soluble in water (100 mM),chloroform (1:2),alcohol (1:1),methanol,and DMSO. | |
| CN(C)CCC=C1C2=CC=CC=C2COC3=CC=CC=C31.Cl | |
| 315.841 | |
| 315.84 |
| C19H22ClNO | |
| UN2811 | |
| doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride | |
| MHNSPTUQQIYJOT-SJDTYFKWSA-N | |
| (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride | |
| 6419921 | |
| Doxepin hydrochloride |
Safety and Handling
GHS H Statement
H301-H361
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H301-H361
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 214-966-8
missing translation for 'rtecsNumber' : HQ4375000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.