Learn More
Lovibond™ DPD 1 Reagent Solution
Lovibond DPD 1 Reagent Solution, Quantity: 15mL, Packaging: 1Each, Green
Brand: Lovibond™ 471020
Code : NEW
Additional Details : CAS Number : 7664-93-9 Weight : 0.01500kg Transport : UN number : 2796 Chem class : 8 Pack group : II
Description
Lovibond DPD 1 Reagent Solution, Quantity: 15mL, Packaging: 1Each, Green
Specifications
Green | |
1 Each | |
15 mL |
Safety and Handling
- DPD 1 reagent solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : sulfuric acid, N, N-Diethyl-1, 4-benzoldiamin-sulfat (1:1)