Learn More
Dräger™ Short-Term Detector Tubes: Vinyl Chloride
Detector tubes
Brand: Dräger™ 8101721
Code : NEW
Additional Details : Weight : 0.07380kg
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Vinyl Chloride | |
5 to 30 | |
10 | |
50°F to 86°F | |
Vinyl chloride 0.5/b |
20mg/L | |
1/5 | |
2.5 min./30 sec. | |
10°C to 30°C |
Safety and Handling
- draeger tubes
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.