missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drierite™, without indicator, 6 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
Specifications
Melting Point | 1450.0°C |
Physical Form | Granular |
Quantity | 500 g |
Packaging | Glass Bottle |
Solubility Information | Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol |
IUPAC Name | calcium sulfate |
Formula Weight | 136.14 |
Chemical Name or Material | Drierite™ |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'