missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DsbA Monoclonal Antibody (8A10)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA518312

Product Code. 15577261

  • 566.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects endogenous levels of DsbA at a molecular weight of 24 kDa and does not cross-react with related proteins. Purity is >95% by SDS-PAGE.

DsbA is required for disulfide bond formation in some periplasmic proteins such as PhoA or OmpA. Acts by transferring its disulfide bond to other proteins and is reduced in the process. DsbA is reoxidized by DsbB. It is required for pilus biogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DsbA
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Mouse
Antigen affinity chromatography
RUO
Bacteria
Antibody
IgG1
Western Blot
8A10
Unconjugated
Disulphide oxidoreductase; Thiol disulfide interchange protein
Recombinant full length DsbA from E. coli.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.