missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuraCal™ Redox Buffer pH 4.01, Hamilton™
Click to view available options
Quantity:
5 L
Specifications
Color | Red |
pH | 4.01 |
Physical Form | Liquid |
Quantity | 5 L |
Solution Type | Buffer |
Odor | Odorless |
Solubility Information | Water Soluble |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'