Learn More
DWK Life Sciences DURAN™ Acid Storage Bottle, with ST 19/17 ground joint and pennyhead glass stopper
DURAN™ clear glass acid storage bottle.
Brand:  DWK Life Sciences 212754409
Code : NEW
Additional Details : Weight : 6.53000kg
 View more versions of this product
Product Code. 15261679
  • 980.00 € / Pack of 10
Estimated Shipment
03-08-2020
Add to basket
Description
Description
Made of chemically resistant DURAN™ glass. With standard taper pennyhead stopper and additional ground glass cap. This provides an improved seal and protection against acid vapours.
  • Ground DURAN™ glass stopper and additional DURAN™ glass cap provides protection against acid vapours
  • Pennyhead stopper with standard taper joint
  • Conical shoulders
  • Replacable glass cap
Specifications
Specifications
Individual Components