Learn More
DWK Life Sciences DURAN™ Filter Disc Sintered Glass, Porosity 2, edge not fused
A prerequisite for successful work with glass filters is the selection of the right porosity. It is important to select a filter whose largest pore size is slightly below the grain size of the smallest particles still to be separated.
14.15 € - 80.35 €
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
10675971
|
DWK Life Sciences
251590208 |
90 mm |
33.20 €
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
15272029
|
DWK Life Sciences
251530202 |
30 mm |
55.40 €
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15222039
|
DWK Life Sciences
251540203 |
40 mm |
72.05 €
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
11610019
|
DWK Life Sciences
251600206 |
100 mm |
70.85 €
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11689988
|
DWK Life Sciences
251545201 |
45 mm |
80.35 €
Pack of 10 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11620009
|
DWK Life Sciences
251565203 |
65 mm |
23.10 €
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
15921501
|
DWK Life Sciences
251535209 |
35 mm |
60.70 €
Pack of 10 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
11649998
|
DWK Life Sciences
251555202 |
55 mm |
14.15 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10101651
|
DWK Life Sciences
251620208 |
120 mm |
79.15 €
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
15232019
|
DWK Life Sciences
251510209 |
10 mm |
47.20 €
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15262039
|
DWK Life Sciences
251550204 |
50 mm |
48.50 €
Pack of 5 |
In Stock
Log in to see stock. |
|||||
10451073
|
DWK Life Sciences
251580207 |
80 mm |
29.35 €
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10283741
|
DWK Life Sciences
251560205 |
60 mm |
15.40 €
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
10519201
|
DWK Life Sciences
251570206 |
70 mm |
26.80 €
Each |
Estimated Shipment: 09-07-2024
Log in to see stock. |
|||||
15272019
|
DWK Life Sciences
251520201 |
20 mm |
49.05 €
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15222029
|
DWK Life Sciences
251525208 |
25 mm |
50.25 €
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15505026
|
DWK Life Sciences
251515207 |
15 mm |
48.70 €
Pack of 10 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
Description
Glass powder with a certain particle size distribution results in a porous, solid body. These bodies are used as filter material in the form of plates and candles. Made of DURAN™ (borosilicate glass 3.3). They are ideally suited for separations with strong acids or alkalis, for example, and thus offer advantages over other materials such as plastic or paper. Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99. Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar.
- Edge non-melted and centered, surfaces untreated
- Successful work through the correct choice of porosity (The largest pore size should be slightly below the grain size of the smallest particles still to be separated).
- Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99.
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |