Learn More
DWK Life Sciences DURAN™ Filter Disc Sintered Glass, Porosity 4, edge not fused
A prerequisite for successful work with glass filters is the selection of the right porosity. It is important to select a filter whose largest pore size is slightly below the grain size of the smallest particles still to be separated.
17.25 € - 105.00 €
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
15292019
|
DWK Life Sciences
251520407 |
20 mm |
55.40 €
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15292029
|
DWK Life Sciences
251530408 |
30 mm |
65.55 €
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15352210
|
DWK Life Sciences
251540409 |
40 mm |
86.75 €
Pack of 10 |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10116440
|
DWK Life Sciences
251590405 |
90 mm |
43.60 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
10441073
|
DWK Life Sciences
251620405 |
120 mm |
105.00 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
10298390
|
DWK Life Sciences
251560402 |
60 mm |
19.25 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11609998
|
DWK Life Sciences
251545407 |
45 mm |
96.95 €
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11679998
|
DWK Life Sciences
251555408 |
55 mm |
17.25 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11640009
|
DWK Life Sciences
251565409 |
65 mm |
28.20 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11630019
|
DWK Life Sciences
251600403 |
100 mm |
91.95 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
15931501
|
DWK Life Sciences
251535406 |
35 mm |
72.05 €
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
10253361
|
DWK Life Sciences
251570403 |
70 mm |
34.60 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
15242019
|
DWK Life Sciences
251510406 |
10 mm |
53.50 €
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15242029
|
DWK Life Sciences
251525405 |
25 mm |
58.10 €
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15282039
|
DWK Life Sciences
251550401 |
50 mm |
56.35 €
Pack of 5 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
10298440
|
DWK Life Sciences
251580404 |
80 mm |
38.30 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11689978
|
DWK Life Sciences
251515404 |
15 mm |
55.40 €
Pack of 10 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
Glass powder with a certain particle size distribution results in a porous, solid body. These bodies are used as filter material in the form of plates and candles. Made of DURAN™ (borosilicate glass 3.3). They are ideally suited for separations with strong acids or alkalis, for example, and thus offer advantages over other materials such as plastic or paper. Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99. Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar.
- Edge non-melted and centered, surfaces untreated
- Successful work through the correct choice of porosity (The largest pore size should be slightly below the grain size of the smallest particles still to be separated).
- Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99.
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |