missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ DURAN™ Test Tubes with Blue Graduations

Manufactured from DURAN™ borosilicate glass 3.3 for excellent chemical and thermal shock resistance.

Brand:  Hirschmann™ 6400165

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 13198019

  • 89.00 € / Pack of 10
Estimated Shipment: 18-07-2024
to see stock.
Add to basket

Description

Description

  • With spout for easy and spill-free pouring
  • Round base
  • Clear glass for excellent clarity
  • Available in a range of capacities
Specifications

Specifications

Clear
20 mL
Borosilicate Glass
10/Pk.
17 mm
Graduation increments of 0.2 mL
205 mm
Spout
Yes
Test Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers