Learn More
DWK Life Sciences DURAN™ Pipeline Filter
A pipeline filter is used to separate solids from a gas phase.
Brand: DWK Life Sciences 258551306
Code : NEW
Additional Details : Weight : 0.15500kg
Description
In addition to solid-liquid filtration, filtration or separation of reaction mixtures from the gas phase is also possible. If, for example, a pipeline filter is installed in a hose system, these filters solid residues from the gas phase and thus purifies it.
- By installing the filter in a pipe system, the purification of a carrier gas is possible
- Fired-on, highly durable white print
Specifications
Porosity 3 | |
60 mm plate | |
Boro 3.3 | |
1/Pk. |