missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Protect Clear Borosilicate Glass Laboratory Bottles with Plastic Safety Coating, DIN Thread, Graduations, Bottle Only
605.64 EUR valid until 2024-05-31
Use promo code "21660" to get your promotional price.
605.64 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
605.64 EUR valid until 2024-12-31
Use promo code "21932" to get your promotional price.
774.77 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "21660" to get your promotional price.
605.64 EUR valid until 2024-06-30
Use promo code "21742" to get your promotional price.
605.64 EUR valid until 2024-12-31
Use promo code "21932" to get your promotional price.
774.77 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Click to view available options
Capacity (Metric):
10 mL
10,000 mL
100 mL
1000 mL
15,000 mL
150 mL
20,000 mL
2000 mL
25 mL
250 mL
3500 mL
50 mL
500 mL
5000 mL
750 mL
Description
- DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972.
- The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside.
- The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
- A combination of high-performance materials and robust ISO 4796-1 design provides a long working life.
- The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel.
- Permanent Retrace Code on each bottle for manufacturing lot traceability.
- The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134°C.
- The coating provides scratch, leak* and splinter* protection (* only applies to bottles 5000 mL and less).
- Highly resistance, long service-life coating.
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out.
- UV protection up to approx. 380 nm due to external coating.
- Easy-to-read scale with graduation marks to estimate the content.
- Fired-on, highly durable white ceramic print.
- Sizes 10 and 25 mL with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
Specifications
Capacity (Metric) | 2000 mL |
Capacity (English) | 0.5 gal. |
Coating | Polyurethane |
Color | Clear |
Diameter (English) | 5.4 in. |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
Includes | Without screw cap and pouring ring |
Material | Borosilicate Glass (3.3), Polypropylene |
Quantity | 10 Pk. |
Thread Size (Metric) | GL 45 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'