Learn More
DWK Life Sciences DURAN™ Tube with two Hose Connections
For attaching to a wide variety of apparatus.
Brand: DWK Life Sciences 286022206
157.70 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 2.25000kg
Description
Hoses can be connected to laboratory apparatus via DURAN™ tube nozzles. There is always a risk of injury due to glass breakage when dismantling tightly fitted hoses from glass olives. DWK therefore recommends the use of DURAN™ glass threads with plastic nozzles.
- Hose olives enable the connection of hoses to laboratory apparatuses
- Glass articles for further processing
Specifications
250/Pk. | |
For attaching to various apparatuses; Installation in different apparatus possible | |
Tube with Connections | |
9 mm |