missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Duratouch™ 34-755 Series Single Use Vinyl Gloves

Provides a stronger, economical alternative to household rubber gloves. Ansell Edmont™ Duratouch™ 34-755 Series Single Use Vinyl Gloves are ambidextrous single-use vinyl gloves.

Brand:  Ansell Edmont™ 34755065

Code : NEW

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15329177

  • 11.50 € / Pack of 100

Estimated Shipment date
from Supplier 02-06-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Single use gloves
  • Reduces costs for hand/ product protection wherever changeover rate is high
  • No seams to leak or split
  • Liquid proof, with no point of weakness
  • Pre-powdered
  • More comfortable to wear and easier to put on and remove without tearing
  • Gloves contain no talcum powder which might contaminate products
  • Ambidextrous, fits either hand
  • More economical and convenient: no need to pair left and right

Applications:

  • Painting, coating and cleaning
  • Laboratory analysis
  • Food Processing
  • Agriculture
  • Maintenance
Specifications

Specifications

Smooth Finish
Vinyl
34-755
0.12mm
Single use gloves
240 mm
Rolled beaded
100 gloves in box, 10 boxes in carton
6.5
Vinyl gloves
Clear
EN 420:2003 + A1:2009 and EN374:2003
Videos
Documents