Learn More
ABEBA™ Dynamic Anthracite Occupational Clogs Ob ESD
Anthracite dynamic occupational clogs with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
Brand:  ABEBA™ 37328/41
Additional Details : Weight : 0.79200kg
 View more versions of this product
Product Code. 17131355
  • 66.20 € / 1 pair
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • Breathable, lightweight, comfortable occupational clogs
  • Comply to EN ISO 20347: 2012, OB SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Water repellent anthracite knitted textile
  • Non foldable heel strap, adjustable
  • Removable active comfort insole
Specifications
Specifications
Anthracite
41
Unisex
Water Resistant
Knitted Fabric
ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC
Clogs
OB SRC ESD